KNF UN026.2ANI EX Explosion Proof Motor Vacuum Pump, Al/NBR; 1.6 cfm/27"Hg-29psi

Representative image only.

KNF UN026.2ANI EX Explosion Proof Motor Vacuum Pump, Al/NBR; 1.6 cfm/27"Hg-29psi

KNF | Mfr: 072943/076071| Item: 0705786

Oversized, permanently lubricated bearings for long-term operation

$3,982.57CAD / Each

$3,982.57CAD / Each

Buy It Together

Current Item

KNF UN026.2ANI EX Explosion Proof Motor Vacuum Pump, Al/NBR; 1.6 cfm/27"Hg-29psi
KNF UN026.2ANI EX Explosion Proof Motor Vacuum Pump, Al/NBR; 1.6 cfm/27"Hg-29psi
0705786

$3,982.57CAD / Each
+ =
Total Price 11887.31 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

More About this Item

Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system.

Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.

1 Year Warranty
Specifications
  • Free-Air Capacity (CFM)1.6
  • Free-Air Capacity (LPM)45
  • Free-Air Capacity (LPH)2700
  • Max Vacuum (in Hg)27
  • Max Vacuum (mbar)100
  • Max Vacuum (mm Hg)74.2
  • Max Vacuum (torr)74.2
  • Port Size1/4" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115 / 230
  • Power (amps)2.5
  • Power (Hz)50/60
  • Height (in)10.9375
  • Width (in)7.25
  • Length (in)13.75
  • Height (cm)27.78
  • Width (cm)18.42
  • Length (cm)34.93
  • Height (mm)277.8
  • Width (mm)184.2
  • Length (mm)349.3
  • DescriptionExplosion Proof Motor Vacuum Pump, Al/NBR; 1.6 cfm/27"Hg-29psi
  • Warranty1 Year
Reviews
Q&A