KleenGuard 97822 Gray Nitrile Gloves, Medium, Box/150
Representative image only.
KleenGuard 97822 Gray Nitrile Gloves, Medium, Box/150
KleenGuard | Mfr: 97822| Item: 8648403
Get 50% more gloves per case, reduces packaging waste
$134.08CAD / Box of 150
BUY MORE & SAVE
  • 1-9$134.08 CAD / Box of 150
  • 10+$120.67 CAD / Box of 150
$134.08CAD / Box of 150
BUY MORE & SAVE
  • 1-9$134.08 CAD / Box of 150
  • 10+$120.67 CAD / Box of 150
Product Description
More About this Item
KleenGuard G10 grey nitrile gloves are latex-free and designed for tasks requiring maximum dexterity and tactile sensitivity. These gloves are also environmentally friendly since you get 50% more gloves per case — resulting in less packaging and added space savings. Gloves are powder-free, nonsterile, and have rolled cuffs. Finger tips are textured for added grip.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • MaterialNitrile
  • Thickness (mil)3.5
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorGray
  • GripTextured
  • CuffRolled
  • SterileNo
  • DescriptionGray Nitrile Gloves, Medium, Box/150