Kimtech HC61190-56894 Sterile Critical Nitrile Gloves, large, bag of 20 pair
Representative image only.
Kimtech HC61190-56894 Sterile Critical Nitrile Gloves, large, bag of 20 pair
Kimtech | Mfr: HC61190-56894| Item: 8648602
Hand specific gloves are individually pair packed
$72.79CAD / Bag of 20
BUY MORE & SAVE
  • 1-9$72.79 CAD / Bag of 20
  • 10+$65.51 CAD / Bag of 20
$72.79CAD / Bag of 20
BUY MORE & SAVE
  • 1-9$72.79 CAD / Bag of 20
  • 10+$65.51 CAD / Bag of 20
Product Description
Key Features
  • Enhanced natural grip
  • Powder-free
More About this Item
Lightly textured, powder-free white nitrile gloves feature a beaded cuff for easier donning. Lightly textured palms and fingertips enhance natural grip. Gloves are hand specific with a winged thumb for better fit and mobility. Gloves are sterilized via gamma irradiation and individually pair packed in a double polybag.
Specifications
  • Thickness (mil)5
  • MaterialNitrile
  • SizeLarge
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • GripTextured
  • ComplianceISO 11137
  • SterileYes
  • DescriptionSterile Critical Nitrile Gloves, large, bag of 20 pair