Kimberly-Clark OL30-06C1.5I Reader Safety Glasses, Clear Lens, Black Frame; +1.5 Diopter
Representative image only.
Kimberly-Clark OL30-06C1.5I Reader Safety Glasses, Clear Lens, Black Frame; +1.5 Diopter
Kimberly-Clark | Mfr: 19878| Item: 8647420
Combines magnification and safety in a pair of glasses
$52.60CAD / Each
BUY MORE & SAVE
  • 1-11$52.60 CAD / Each
  • 12+$47.34 CAD / Each
$52.60CAD / Each
BUY MORE & SAVE
  • 1-11$52.60 CAD / Each
  • 12+$47.34 CAD / Each
Product Description
Key Features
  • Nylon frame with 7-base lens for close coverage around eyes
More About this Item
Solves the problem of workers removing safety glasses to put on reading glasses throughout the workday. Precise placement of lens insert, like a bifocal, reduces headaches and eyestrain. Compliance: ANSI Z87.1-2003
Specifications
  • Frame ColorBlack
  • Lens ColorClear
  • ComplianceANSI Z87.1-2003
  • Magnification+1.5
  • DescriptionReader Safety Glasses, Clear Lens, Black Frame; +1.5 Diopter