ITS 480115 Water Quality Test Strips, Multi-Test Strip.

Representative image only.

ITS 480115 Water Quality Test Strips, Multi-Test Strip.

ITS | Mfr: 480115| Item: 9953115

Obtain test results within 90 seconds

$49.51CAD / Pkg of 30

$49.51CAD / Pkg of 30
Product Description

Key Features

  • Test strips feature bright easy-to-read colors
  • No hazardous reagents

More About this Item

These economical water quality test strips are formulated to test tap water, lake or stream water, rain run-off water, or treated water.

The multi-test strips come complete with tests for:
Free chlorine: 0, 0.05, 0.1, 0.2, 0.4, 0.5, 0.8, 1.0, 2.0, 5.0 ppm
Total chlorine: 0, 0.05, 0.075, 0.1, 0.125, 0.15, 0.175, 0.2, 0.3, 0.5, 0.8, 4.0, 10.0 ppm
pH: 6.0, 6.5, 7.0, 7.5, 8.0, 8.5, 9.0
Total alkalinity: 0, 80, 120, 180, 240, 360
Total hardness: 0, 50, 120, 250, 425 ppm

Specifications
  • TestpH, free and total chlorine, hardness, alkalinity
  • Measurement PointsFree chlorine, Total chlorine, pH, Total alkalinity and, Total hardness
  • Reagent PlatformStrips
  • Tests Per Kit1
  • DescriptionWater Quality Test Strips, Multi-Test Strip.
Reviews
Q&A