International Products Corp Surface-Cleanse/930 Liquid Concentrate; 200 kg Drum
Representative image only.
International Products Corp Surface-Cleanse/930 Liquid Concentrate; 200 kg Drum
International Products Corp | Mfr: S-2090-055| Item: 1810013
Easily clean glass, metal, plastic, rubber, and other hard materials
$8,184.68CAD / Each
$8,184.68CAD / Each
Product Description
Key Features
  • Leaves no charged residue—perfect for cleaning electrical components!
More About this Item
Safely wash away grease, petroleum, fingerprints, oils, scale, oxides, and metallic residues from glass, aluminum, nickel, lead, zinc, ceramic, and plastic without surface damages typically caused by acidic or alkaline cleaners. Use to clean filter membranes and other delicate items. Non-flammable and non-hazardous under U.S. federal regulations for shipping, storage, and disposal. Meets USDA guidelines for incidental contact with food processing systems. Contains no CFCs, phosphates, silicates, borates, halogens, or phenols.
Specifications
  • FormatLiquid
  • Dilution1:100 to 2:100, pH 6.5
  • ApplicationsEmulsifiers, Gels, Oils, Fragrances, Petrolatum, Paraffins
  • DescriptionSurface-Cleanse/930 Liquid Concentrate; 200 kg Drum