IC Biomedical R033-8C00 Roller Base for Dewars with 18.8" dia Base
Representative image only.
IC Biomedical R033-8C00 Roller Base for Dewars with 18.8" dia Base
IC Biomedical | Mfr: R033-8C00| Item: 0377371
$781.63CAD / Each
$781.63CAD / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Easily move storage Dewars and Dewars with canister storage systems. Sturdy cast aluminum frame has five wheels with ball-bearing swivel casters. Use model 03773-65 for Dewars with 15.6" diameter base; use 03773-71 for Dewars with 18.8" diameter base. Shpg wt 8.0 lb (3.7 kg). Note: Roller base does not include tipping stand.
Two year warranty
Specifications
  • DescriptionRoller Base for Dewars with 18.8" dia Base