Horiba OCMA-550-100-240V Oil Content Analyzer 100 -240VAC

Representative image only.

Horiba OCMA-550-100-240V Oil Content Analyzer 100 -240VAC

Horiba | Mfr: 3200577566| Item: 8290005

Routine analysis of oil residuals

$19,333.82CAD / Each

$19,333.82CAD / Each

Buy It Together

Current Item

Horiba OCMA-550-100-240V Oil Content Analyzer 100 -240VAC
Horiba OCMA-550-100-240V Oil Content Analyzer 100 -240VAC
8290005

$19,333.82CAD / Each
+ =
Total Price 20989.91 ADD 2 ITEMS TO CART
Product Description

Includes

Quartz cell with cap, power cord, B-heavy oil (standard), and 2 amp fuse.

Key Features

  • Easily measure oil in water or soil for a wide
  • 1 year warranty

More About this Item

The Horiba OCMA-550 analyzer simplifies measurement of oil in water or soil for a wide variety of applications. Use this precision instrument for surveying water quality and hazardous waste sites, industrial wastewater monitoring, post-cleaning discharge analysis, and measuring adherent oil residuals on textiles and metal parts.

Use S-316 solvent to quantitatively extract sample. The oil / solvent solution is then simply transferred to the OCMA-550 analyzer for single-button measurement. Analysis is based on infrared (IR) spectral data, with peak integration (at approximately 3.4 µm) to determine oil concentration. S-316 solvent is non-absorbent at this IR wavelength, so only oil is measured. Accurately measures oil down to 0.1 ppm. Auto-calibrates after initial standard reading.

Use built-in time stamping to document results. Read results on LCD or transfer data using parallel printer port or to computer via RS-232. S-316 solvent can be recycled using Solvent Reclaimer SR-300 (82900-20), sold separately below.

1 Year Warranty
Specifications
  • Photometric Readout TypesAbsorbance, concentration
  • Pathlength (mm)200 mm
  • Min Photometric - Absorbance (A)0
  • Max Photometric - Absorbance (A)1
  • DetectorPyroelectric
  • OutputRS-232
  • Display TypeLCD
  • Width (cm)25
  • Height (cm)20
  • Depth (cm)28.5
  • Width (in)9 51/64
  • Height (in)7 29/32
  • Depth (in)11 1/5
  • Power (VAC)100 to 240
  • DescriptionOil Content Analyzer, 100 -240 VAC
  • Warranty1 year
Reviews
Q&A