Honeywell Chart Paper, 10", 0-100, 7 Day; 100/Pk

Representative image only.

Honeywell Chart Paper, 10", 0-100, 7 Day; 100/Pk

Honeywell | Mfr: 24001661-001| Item: 8066180

Replacement paper for your Honeywell chart recorder

$132.75CAD / Pkg of 100

$132.75CAD / Pkg of 100

Buy It Together

Current Item

Honeywell Chart Paper, 10", 0-100, 7 Day; 100/Pk
Honeywell Chart Paper, 10", 0-100, 7 Day; 100/Pk
8066180

$132.75CAD / Pkg of 100
+ +
Honeywell Replacement Chart Recorder Thermal Pen
8066392

$775.00CAD / Each
=
Total Price 11309.88 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Varying temperature ranges and speeds
  • 18 month warranty

More About this Item

Choose the paper compatible with your chart recorder and application needs.

18 Month Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)10
  • DescriptionChart Paper, 10", 0-100, 7 Day; 100/Pk
  • Warranty18 months
Reviews
Q&A