Honeywell 54001 North Full-Face Respirator, Thermoplastic elastomer
Representative image only.
Honeywell 54001 North Full-Face Respirator, Thermoplastic elastomer
Honeywell | Mfr: 54001| Item: 8160622
Wraparound lens provides over 200° field of vision
$355.82CAD / Each
$355.82CAD / Each
Buy It Together
Current Item
Honeywell 54001 North Full-Face Respirator, Thermoplastic elastomer
Honeywell 54001 North Full-Face Respirator, Thermoplastic elastomer
8160622
$355.82CAD / Each
+ =
Total Price 430.67 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Superior comfort and fit
  • Contoured flange ensures proper seal
More About this Item
Soft, pliable silicone and economical thermoplastic elastomer models, both feature hard-coated, scratch-and impact-resistant wraparound polycarbonate lens providing over 200° field of vision. Lens protects eyes and face against irritating gases, vapors and flying particles. The oral/nasal cup with dual sealing flange reduces fogging. Silicone face piece features a speaking diaphragm for clear communication.
Order filter cartridges according to demands of work environment (cartridges sold separately).
Compliance: ANSI Z87.1-1989
Approval: NIOSH
Specifications
  • ComplianceNIOSH; ANSI Z87.1-1989
  • ColorBlack
  • DescriptionFull-Face Respirator, Thermoplastic elastomer