Honeywell 11380048 North Adjustable Head Gear, Black HDPE
Representative image only.
Honeywell 11380048 North Adjustable Head Gear, Black HDPE
Honeywell | Mfr: 11380048| Item: 8169000
Protect yourself against liquid splashes, sparks, and impacts
$44.05CAD / Each
BUY MORE & SAVE
  • 1-9$44.05 CAD / Each
  • 10+$39.65 CAD / Each
$44.05CAD / Each
BUY MORE & SAVE
  • 1-9$44.05 CAD / Each
  • 10+$39.65 CAD / Each
Buy It Together
Current Item
Honeywell 11380048 North Adjustable Head Gear, Black HDPE
Honeywell 11380048 North Adjustable Head Gear, Black HDPE
8169000
$44.05CAD / Each
+ =
Total Price 75.73 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Sweatband absorbs perspiration
  • Can be used with earmuffs
  • ANSI Z87.1, CSA Z94.3 compliance
  • Adjustable fit for tension and visor position
  • Splash drip edge redirects splashes
  • Different shield options (Face Shield Sold Separately)
  • 1 year warranty
More About this Item
SnapLok™Head Gear features adjustable ratchet suspension and tension knob for precise sizing and visor positioning. Splash drip edge redirects splashing or spraying fluids.
Universal Shields are extra wide for excellent protection. Lightweight construction provides comfort during lengthy work periods. Shields attach easily and securely to head gear—no snaps, posts, or levers.
Model 81690-10 is heat-resistant to temperatures up to 220°>F; model 81690-20 withstands temperatures up to 280°>F. Order shields separately.
1 Year WarrantyCanadian Standards Association
Specifications
  • Frame ColorBlack
  • Lens ColorClear
  • ComplianceANSI Z87.1; CSA Z94.3
  • DescriptionAdjustable Head Gear, Black HDPE