Hirschmann opus Titrator, 20 mL; 100 to 240 VAC, 50/60 Hz

Representative image only.

Hirschmann opus Titrator, 20 mL; 100 to 240 VAC, 50/60 Hz

Hirschmann | Mfr: 9582020| Item: 2550305

Obtain precise titration results with electronic control and dispensing

$5,370.51CAD / Each

$5,370.51CAD / Each
Product Description

Includes

opus titrator with A45 thread, additional A32, A38, and S40 thread adapters, suction hose with union nut, recirculation hose, titration ejection unit, valve fitting wrench, touchscreen control unit, light protection window, data cable, power supply unit, manual, and quality certificate.

Key Features

  • Configure titration parameters on user-friendly TFT touch screen
  • Time-saving rapid titration in conjunction with fine titration ensures system accuracy
  • Save up to nine procedures and settings with on-board memory for quick recall
  • Easily transfer data via RS-232 and USB interfaces
  • Titration range from 0.1 to 20 mL
  • Velocity rate in 10 stages from 0.1 mL/s to 4 mL/s
  • 1 year warranty

More About this Item

The electronic titration system is user-friendly with an intuitive touch screen and direct titration from the laboratory bottle. Titration impulse triggered by the motor allows for droplet titration of 1 droplet per button push. Prevent reagent loss with simple and rapid venting, made possible with the media recirculating system.
Advanced safety and protection features are valuable for daily routines. Protect your data and procedures from unauthorized personnel using the PIN function. The PIN function can also enable only select functions to be performed by selected users. Freely rotate the titrator on the reagent bottle, to ensure the bottle label is always pointing towards the user. Your light-sensitive samples will be protected by the light protection window, included with your titration system.

1 Year Warranty
Specifications
  • Display Type3.5" TFT Touchscreen
  • Capacity (mL)20
  • Dispensing AccuracyR=0.2%; CV=0.07%
  • OutputRS-232 and USB
  • Data LoggingUp to 9 configurations
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Length (in)5 1/10
  • Width (in)4 19/64
  • Height (in)2 1/5
  • Length (cm)13
  • Width (cm)11
  • Height (cm)5.5
  • DescriptionTitrator, 20 mL; 100 to 240 VAC, 50/60 Hz
  • Warranty1 year
Reviews
Q&A