Hirschmann 934200010 Ceramus Adjustable-Volume Dispenser, 2.0 to 10.0 mL

Representative image only.

Hirschmann 934200010 Ceramus Adjustable-Volume Dispenser, 2.0 to 10.0 mL

Hirschmann | Mfr: 934200010| Item: 2520736

$1,079.99CAD / Each

for an additional
$155.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$1,079.99CAD / Each

Buy It Together

Current Item

Hirschmann 934200010 Ceramus Adjustable-Volume Dispenser, 2.0 to 10.0 mL
Hirschmann 934200010 Ceramus Adjustable-Volume Dispenser, 2.0 to 10.0 mL
2520736

$1,079.99CAD / Each
+ =
Total Price 1235.43 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Recirculation system allows fast priming without loss of reagent — environmentally safe and economical
  • 1 year warranty

More About this Item

Use these new dispensers in a wide temperature range from 4 to 50°C. The ergonomically designed grip makes dispensers safe and easy to use. Step gauge guarantees precise reproducibility. The repeated setting of a previously set volume is always identical — no calibration is necessary. The exact click-stop latching mechanism ensures an accurate setting of the desired volume.
Dispensers rotate 360° on the bottle so you can easily position the spout and label where you want it for each application. Microstep volume adjustment is calibrated for the lifetime of the device. Steam sterilize dispensers at 121°C without dismantling.
Dispensers feature a Luer-lock rear connection for a filter or dry tube. The ceramic piston gives you excellent chemical resistance without abrasion or swelling. Individual test certificates are included indicating the test values as well as the individual serial number. Complies with DIN EN ISO 9000.

1 Year Warranty
Specifications
  • Min Dispense Volume (mL)2
  • Max Dispense Volume (mL)10
  • Division (mL)0.25
  • Accuracy±0.5%
  • Reproducibility±0.1%
  • AutoclavableYes
  • Wetted PartsCylinder, valves, ball and seat, borosilicate glass, Hastelloy C® spring, and AL2O3 piston
  • DescriptionCeramus Adjustable-Volume Dispenser, 2.0 to 10.0 mL
  • Warranty1 year
Reviews
Q&A