HF Scientific 10478 Chlorine Pocket Colorimeter

Representative image only.

HF Scientific 10478 Chlorine Pocket Colorimeter

HF Scientific | Mfr: 10478| Item: 9956130

Laboratory accuracy in the field

$1,233.01CAD / Each

$1,233.01CAD / Each

Buy It Together

Current Item

HF Scientific 10478 Chlorine Pocket Colorimeter
HF Scientific 10478 Chlorine Pocket Colorimeter
9956130

$1,233.01CAD / Each
+ +
Environmental Express Model 200 Chlorine Colorimeter
9956178

$1,280.38CAD / Each
=
Total Price 2574.39 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

four measuring cuvettes with caps, 4 AAA batteries, and carrying case

Key Features

  • Simple operation
  • Waterproof
  • EPA approved method

More About this Item

This handheld colorimeter provides accuracy expected in the laboratory no matter where you happen to be. The extra- large LCD is easy to read and the auto shut off feature conserves battery life.

DPD reagents are not included in the kit to allow you to choose the type and quantity needed for your application. Reference standard solution is available for calibration.

IP 67
Specifications
  • Display TypeLCD
  • Light SourceLED
  • Wavelength(s) (nm)515
  • Test ParametersChlorine
  • WaterproofYes
  • Battery4 AAA (included)
  • Length (in)6 1/2
  • Width (in)2 3/4
  • Height (in)1 7/8
  • DescriptionChlorine Pocket Colorimeter
Reviews
Q&A