Heraeus Noblelight Hollow Cathode Lamp, chromium, 50 mm, Perkin Elmer Lumina coded
Representative image only.
Heraeus Noblelight Hollow Cathode Lamp, chromium, 50 mm, Perkin Elmer Lumina coded
Heraeus Noblelight | Item: 1194986
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
$688.81CAD / Pkg of 1
$688.81CAD / Pkg of 1
Product Description
Key Features
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
More About this Item
Heraeus Noblelight chromium (Cr) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.
Specifications
  • MaterialChromium
  • Compatible WithPerkin Elmer Lumina
  • Description Hollow Cathode Lamp, chromium, 50 mm, Perkin Elmer Lumina coded