HD Electric DVM-80UVM Dual Stick Digital Voltmeter, True RMS, 0-40 kV
Representative image only.
HD Electric DVM-80UVM Dual Stick Digital Voltmeter, True RMS, 0-40 kV
HD Electric | Mfr: DVM-80UVM| Item: 2302249
Get safe and accurate voltage readings
$4,287.56CAD / Each
for an additional
$490.64CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$4,287.56CAD / Each
Buy It Together
Current Item
HD Electric DVM-80UVM Dual Stick Digital Voltmeter, True RMS, 0-40 kV
HD Electric DVM-80UVM Dual Stick Digital Voltmeter, True RMS, 0-40 kV
2302249
$4,287.56CAD / Each
+ +
HD Electric S-6H 6 ft Extension Hotstick
2002199
$552.52CAD / Each
=
Total Price 5330.72 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Feature a backlit display for low-light conditions, auto shut-off, and low-battery indicator
  • Powered by 9 V batteries
  • True RMS voltage measurements
  • 1 year warranty
More About this Item
The DigiVolt® series of digital voltmeters/phasing sets are designed to take voltage measurements and determine if two conductors are in-phase or out-of-phase on system voltages.
The DVM-80UVM is a compact, high accuracy dual-stick voltmeter and phasing set for virtually all overhead and underground applications up to 40 kV. This is the same as the DVM-80, but with the added features of capacitive test point and peak hold modes.
†Dual-stick meters and kit will require two hotsticks; one for each probe.
1 Year Warranty
Specifications
  • DC Min (Volts)0
  • DC Max (Volts)40000
  • AC Min (Volts)0
  • AC Max (Volts)40
  • DescriptionDual Stick Digital Voltmeter, True RMS, 0-40 kV