Hamilton PRP-1 Reverse Phase Column, High Resolution, 150x4.1mm ID; 5um

Representative image only.

Hamilton PRP-1 Reverse Phase Column, High Resolution, 150x4.1mm ID; 5um

Hamilton | Mfr: 79444| Item: 0194100

High capacity and rapid flow rates for fast analysis

$1,102.10CAD / Each

$1,102.10CAD / Each
Product Description

Key Features

  • Compatible with any concentration of organic solvent
  • 18 month warranty

More About this Item

Hamilton polymeric PRP™ columns are resin-based, offering excellent resolution due to their high selectivity, efficiency, and capacity. The chemically inert macroporous poly(styrenedivinylbenzene)stationary phase is stable from pH 1 to 13. Compatible with any concentration of organic solvent; can be used up to 60°C and 5000 psi. Hamilton guarantees that the spherical media will not shrink or swell.
PRP-1 Reverse Phase Columns contain poly(styrenedivinylbenzene). Large surface area (415 m2/g) allows high capacity and rapid flow rates for fast analysis. Ideal for separating strong acids and bases. Efficiency of 30,000 to 50,000 plates/meter.

18 Month Warranty
Specifications
  • PhasePolystyrenedivinylbenzene
  • Column Length (mm)150
  • Column Inside Diameter (ID) (mm)4.1
  • Column Particle Size (µm)5
  • Wetted MaterialsStainless steel, PSDVB
  • Max Pressure (PSI)5000
  • Min Operating Temperature (° C)5
  • Max Operating Temperature (° C)85
  • ApplicationsOrganic compounds: small molecules (< 2,000 mw), oligonucleotides, pharmaceuticals, steroids, nucleic acids, vitamins, herbicides
  • MethodologyReversed-phase PRP-1
  • DescriptionPRP-1 Reverse Phase Column, High Resolution, 150x4.1mm ID; 5um
  • Warranty18 months
Reviews
Q&A