Hach RD1P5 Differential ORP electrode, 15-ft cable, PEEK body
Representative image only.
Hach RD1P5 Differential ORP electrode, 15-ft cable, PEEK body
Hach | Mfr: RD1P5| Item: 2740014
Unsurpassed accuracy with differential measurement technique
$1,999.39CAD / Each
$1,999.39CAD / Each
Buy It Together
Current Item
Hach RD1P5 Differential ORP electrode, 15-ft cable, PEEK body
Hach RD1P5 Differential ORP electrode, 15-ft cable, PEEK body
2740014
$1,999.39CAD / Each
+ =
Total Price 2187.98 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Three electrodes instead of the normal two in conventional pH sensors
  • Greater reliability resulting in less downtime and maintenance
  • Replaceable salt bridge/protector
  • Encapsulated electronics and preamp
  • Versatile body styles and mounting hardware including retrofit
  • 914 m (3000 ft) transmission distance
  • 1 year warranty
More About this Item
Differential Electrode measurement technique uses three electrodes instead of the two normally used in conventional pH sensors. Process and reference electrodes measure the pH differentially with respect to a third ground electrode. The end result is unsurpassed measurement accuracy, reduced reference junction potential, and elimination of sensor ground loops. These sensors provide greater reliability, resulting in less downtime and maintenance. The unique, replaceable salt bridge holds an extraordinary volume of buffer to extend the working life of the sensor by protecting the reference electrode from harsh process conditions. The salt bridge simply threads onto the end of the sensor if replacement is needed. Encapsulated design of the preamp protects the sensor’s built-in preamp from moisture and humidity, ensuring reliable sensor operation. The preamp in the pH sensor produces a strong signal, enabling the sensor to be located up to 914 m (3000 ft.) from the analyzer.
1 Year Warranty
Specifications
  • Electrode Body MaterialPEEK
  • Min Temperature (° C)-5
  • Max Temperature (° C)95
  • Temperature CompensationYes
  • Max Pressure (PSI)100
  • ATC ConnectorTinned ends
  • ATC ElementNTC 300 Ω thermistor
  • ConnectorTinned ends
  • DescriptionDifferential ORP electrode, 15-ft cable, PEEK body