Hach PO-19A O-Phosphate Color Disc Test Kit , Turbid Samples, Range 0- 0.8, 4, 40 ppm

Representative image only.

Hach PO-19A O-Phosphate Color Disc Test Kit , Turbid Samples, Range 0- 0.8, 4, 40 ppm

Hach | Mfr: 224801| Item: 0554555

Color wheel method provides accurate visual comparisons

$369.82CAD / Each

$369.82CAD / Each

Buy It Together

Current Item

Hach PO-19A O-Phosphate Color Disc Test Kit , Turbid Samples, Range 0- 0.8, 4, 40 ppm
Hach PO-19A O-Phosphate Color Disc Test Kit , Turbid Samples, Range 0- 0.8, 4, 40 ppm
0554555

$369.82CAD / Each
+ + =
Total Price 1531.07 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty

More About this Item

Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.

Includes two reagents, color disc, comparator box, two viewing tubes, viewing adaptor, two bottles, dropper tube, funnel, filter paper for filtration of turbid samples, deionized water, instructions and carrying case.

1 Year Warranty
Specifications
  • Test Phosphorus, Orthophosphate (Reactive)
  • Measurement Points0 to 0.8 ppm; 0 to 4 ppm; 0 to 40 ppm
  • Reagent PlatformColor Disc
  • Tests Per Kit100
  • DescriptionO-Phosphate Color Disc Test Kit , Turbid Samples, Range 0- 0.8, 4, 40 ppm
  • Warranty1 year
Reviews
Q&A