Hach LZW5051T.97.002 pH Electrode, Double-junction for Portable meter
Representative image only.
Hach LZW5051T.97.002 pH Electrode, Double-junction for Portable meter
Hach | Mfr: LZW5051T.97.002| Item: 5900452
A variety of electrodes for different applications
$1,000.43CAD / Each
$1,000.43CAD / Each
Product Description
Key Features
  • Different body matierals for chemical compatibility
  • Glass body, solid polymer filled electrode for dirty water samples
  • Polypropylene shroud offers protection
  • Heavy-duty electrode handle optimized for field calibration and storage
  • Replacement for 59004-03 wastewater kit
  • 6 month warranty
More About this Item
Choose the electrode that is most in line with the pH range of your application.
6 Month Warranty
Specifications
  • ApplicationsSamples with high solids content
  • Electrode TypeOpen annular junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteNon-refillable solid polymer
  • Max pH (pH)14
  • Accuracy0.02 pH with pH1 Meter
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorSension+ MP5
  • ATC Element1000Ω Pt RTD
  • ConnectorSension+ MP5
  • Diameter (mm)12
  • Length (mm)85
  • Cable Length (m)1
  • DescriptionpH Electrode w/ ATC for Portable Meters, Sealed, Solid Polymer Filled, Glass, Open Annular Junction
  • Warranty6 months