Hach LZW5050T.97.002 pH Electrode w/ ATC for Portable Meters, Sealed, Gel-Filled, PC, Ceramic Pin Junction
Representative image only.
Hach LZW5050T.97.002 pH Electrode w/ ATC for Portable Meters, Sealed, Gel-Filled, PC, Ceramic Pin Junction
Hach | Mfr: LZW5050T.97.002| Item: 5900451
A variety of electrodes for different applications
$493.59CAD / Each
$493.59CAD / Each
Product Description
Key Features
  • Different body matierals for chemical compatibility
  • Polycarbonate body, single-junction electrode for clean water samples
  • Storage protector prevents breakage
  • Heavy-duty electrode handle optimized for field calibration and storage
  • Replacement electrode for 59004-01 drinking water kit
  • 6 month warranty
More About this Item
Choose the electrode that is most in line with the pH range of your application.
6 Month Warranty
Specifications
  • ApplicationsGeneral laboratory applications
  • Electrode TypeCeramic pin
  • Electrode Body MaterialPolycarbonate
  • Reference ElectrolyteNon-refillable gel
  • Min pH (pH)0
  • Max pH (pH)14
  • Accuracy0.02 pH with pH1 Meter
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorSension+ MP5
  • ATC Element1000Ω Pt RTD
  • ConnectorSension+ MP5
  • Diameter (mm)12
  • Length (mm)85
  • Cable Length (m)1
  • DescriptionpH Electrode w/ ATC for Portable Meters, Sealed, Gel-Filled, PC, Ceramic Pin Junction
  • Warranty6 months