Hach LTV082.53.40001 DRB 200 COD Reactor, single block, 15 X 16 mm vial wells, 115 VAC
Representative image only.
Hach LTV082.53.40001 DRB 200 COD Reactor, single block, 15 X 16 mm vial wells, 115 VAC
Hach | Mfr: LTV082.53.40001| Item: 9957460
Ideal for wastewater influent and effluent testing, industrial process water testing, and environmental monitoring
$2,616.74CAD / Each
$2,616.74CAD / Each
Product Description
Key Features
  • Uses EPA-approved COD method—requires less equipment, lab space, and maintenance than other open reflux COD tests
  • High flexibility, customization, and temperature accuracy
  • For a complete system order COD reactor (99574-60 or -65), COD or TOC test kits (99574-25,-26,-27,-28, or -29), and colorimeter (99573-00)
  • 2 year warranty
More About this Item
Hach DRB-200 COD Reactor digests up to 15 samples simultaneously. Reactors feature one-touch operation through the use of preprogrammed Hach COD and TNT standard digestion temperatures/times, or customize up to three programs. To customize, choose from one of nine languages, set the temperature between 37 to 165°C, and enter the time between 1‑minute to 8 hours. Built-in timer features audible alarm and automatic shut-off to prevent overheating. Transparent, locking lid covers 16 x 100-mm vials while protecting the user in the unlikely event of a broken reagent vial.
For full specifications and operating instructions view the instruction manual linked below.
UL Listed.Two year warrantyEPA United States Environmental protection agency
Specifications
  • Meter TypeAccessory; Reactors
  • Power (VAC)115
  • DescriptionDRB 200 COD Reactor, single block, 15 X 16 mm vial wells, 115 VAC