Hach HQ4100 Portable Multiparameter pH/Conductivity/TDS/Salinity/DO/ORP/ISE, 1 Channel

Representative image only.

Hach HQ4100 Portable Multiparameter pH/Conductivity/TDS/Salinity/DO/ORP/ISE, 1 Channel

Hach | Mfr: LEV015.53.4100A| Item: 5900528

A robust and intuitive multiparameter meter

$2,705.14CAD / Each

$2,705.14CAD / Each

Buy It Together

Current Item

Hach HQ4100 Portable Multiparameter pH/Conductivity/TDS/Salinity/DO/ORP/ISE, 1 Channel
Hach HQ4100 Portable Multiparameter pH/Conductivity/TDS/Salinity/DO/ORP/ISE, 1 Channel
5900528

$2,705.14CAD / Each
+
Hach HQ USB Cable for Portable HQ Series Meter
5900546

$59.89CAD / Each
=
Total Price 2765.03 ADD 2 ITEMS TO CART
Product Description

Includes

Meter, rechargeable lithium-ion battery, USB power adapter, and USB cable.

Key Features

  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 100,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
  • 3 year warranty

More About this Item

Hach HQ meters offer a versatile range of measurements, including pH, conductivity, total dissolved solids (TDS), salinity, dissolved oxygen (DO), oxidation reaction potential (ORP), ion selective electrode (ISE), and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.

The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 640 x 480 color TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.

3 Year WarrantyConformite Europeenne
Specifications
  • Meter TypepH, conductivity, TDS, salinity, DO, ORP, ISE
  • Display type640x480 Color TFT
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.001/0.01/0.1
  • pH AccuracyProbe specific
  • Buffer RecognitionIUPAC, USA, DIN, Custom
  • CalibrationUp to 3 points
  • Min mV (mV)-1200
  • Max mV (mV)1200
  • mV Resolution0.1 mV
  • mV AccuracyProbe specific
  • Min ISEProbe specific
  • Max ISEProbe specific
  • ISE ResolutionProbe specific
  • ISE AccuracyProbe specific
  • Min Conductivity0.01 µS/cm
  • Max Conductivity200 mS/cm
  • Conductivity Resolution0.01 µS/cm to 0.1 mS/cm upon selected measuring range
  • Conductivity AccuracyProbe specific
  • Min TDS0.01 g/L
  • Max TDS50 g/L
  • TDS Resolution0.01 mg/L up - 0.1 g/L (range dependent)
  • TDS AccuracyProbe specific
  • Min Salinity42 ppt or %
  • Max Salinity0 ppt or %
  • Salinity Resolution0.01 (range dependent)
  • Salinity AccuracyProbe specific
  • Min Dissolved Oxygen0.0 mg/L
  • Max Dissolved Oxygen37 mg/L
  • Dissolved Oxygen Resolution0.1 mg/L
  • Dissolved Oxygen AccuracyProbe specific
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature Resolution 0.1°C
  • Temperature AccuracyProbe specific
  • OutputUSB
  • Data Logging (points)Yes
  • Memory Size100,000 data points
  • WaterproofIP67
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Battery1 rechargeable Lithium-ion (included)
  • Length (cm)22
  • Width (cm)9.7
  • Height (cm)6.3
  • Width (in)3 13/16
  • Length (in)8 21/32
  • Height (in)2 31/64
  • Number of Positions1
  • NIST Calibration IncludedNo
  • Probe IncludedNo
  • DescriptionPortable Multi-Meter pH/Conductivity/TDS/Salinity/DO/ORP/ISE, 1 Channel
  • Warranty3 year
Reviews
Q&A