Hach HQ1140 Portable Dedicated Conductivity/TDS Meter with Conductivity Electrode, 1 m Cable

Representative image only.

Hach HQ1140 Portable Dedicated Conductivity/TDS Meter with Conductivity Electrode, 1 m Cable

Hach | Mfr: LEV015.53.11401| Item: 5900509

A robust and intuitive conductivity and TDS meter

$3,067.60CAD / Each

$3,067.60CAD / Each

Buy It Together

Current Item

Hach HQ1140  Portable Dedicated Conductivity/TDS Meter with Conductivity Electrode, 1 m Cable
Hach HQ1140 Portable Dedicated Conductivity/TDS Meter with Conductivity Electrode, 1 m Cable
5900509

$3,067.60CAD / Each
+
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter
5900500

$1,351.10CAD / Each
+ + =
Total Price 6709.82 ADD 4 ITEMS TO CART
Product Description

Includes

Meter, CDC40101 conductivity cell (59004-63), rechargeable battery, USB power adapter, and USB cable.

Key Features

  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 5,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
  • 1 m cable 4-poles graphite conductivity cell
  • 1 year warranty

More About this Item

Hach HQ portable meters offer a versatile range of measurements, including salinity, total dissolved solids (TDS), and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.

The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.

1 Year WarrantyConformite Europeenne
Specifications
  • Meter TypeConductivity, TDS
  • Display type536x336 Mono-TFT
  • Buffer RecognitionIUPAC, USA, DIN, Custom
  • CalibrationUp to 3 points
  • Min Conductivity0.01 µS/cm
  • Max Conductivity200 mS/cm
  • Conductivity Resolution0.01 µS/cm to 0.1 mS/cm upon selected measuring range
  • Conductivity Accuracy±0.5% of range
  • Min TDS0.01 g/L
  • Max TDS50 g/L
  • TDS Resolution0.01 mg/L up - 0.1 g/L (range dependent)
  • TDS Accuracy±0.5% ±1 digit
  • Salinity Accuracy±0.1, ±1 digit
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature Resolution 0.1°C
  • Temperature Accuracy±0.3 °C (±0.54 °F)
  • Temperature compensationATC
  • OutputUSB
  • Data Logging (points)Yes
  • Memory Size5,000 data points
  • WaterproofIP67
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Battery1 rechargeable lithium-ion (included)
  • Length (cm)22
  • Width (cm)9.7
  • Height (cm)6.3
  • Width (in)3 13/16
  • Length (in)8 21/32
  • Height (in)2 31/64
  • Number of Positions1
  • NIST Calibration IncludedNo
  • Probe Included1 m cable conductivity electrode
  • DescriptionPortable Dedicated Conductivity/TDS Meter with Conductivity Electrode, 1 m Cable
  • Warranty1 year
Reviews
Q&A