Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable

Representative image only.

Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable

Hach | Mfr: LEV015.53.11104| Item: 5900501

A robust and intuitive pH, ORP, and mV meter

$3,561.18CAD / Each

$3,561.18CAD / Each

Buy It Together

Current Item

Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable
Hach HQ1110 Portable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable
5900501

$3,561.18CAD / Each
+ +
Hach HQ1140  Portable Dedicated Conductivity/TDS Meter
5900508

$1,442.45CAD / Each
+ + + +
Hach LDO10101 IntelliCAL LDO Lab Probe, 1 m cable
5900457

$1,794.59CAD / Each
=
Total Price 15187.70 ADD 7 ITEMS TO CART
Product Description

Includes

Meter, MTC101 rugged ORP electrode (59004-66), rechargeable lithium-ion battery, USB power adapter, and USB cable.

Key Features

  • IntelliCal™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures
  • Store up to 5,000 data points
  • Rugged, IP67-rated waterproof housing
  • Option to select from 27 languages in set up mode
  • 1 m low maintenance gel pH electrode
  • 1 year warranty

More About this Item

Hach HQ portable meters offer a versatile range of measurements, including pH, ORP, mV, and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.

The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.

1 Year WarrantyConformite Europeenne
Specifications
  • Meter TypepH, ORP, mV
  • Display type536x336 Mono-TFT
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.001/0.01/0.1
  • pH AccuracyProbe specific
  • Buffer RecognitionIUPAC, USA, DIN, Custom
  • CalibrationUp to 3 points
  • Min mV (mV)-1200
  • Max mV (mV)1200
  • mV Resolution0.1 mV
  • mV Accuracy±0.02 mV or 0.05%, whichever is greater
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature Resolution 0.1°C
  • Temperature Accuracy±0.3 °C (±0.54 °F)
  • Temperature compensationATC
  • OutputUSB
  • Data Logging (points)Yes
  • Memory Size5,000 data points
  • WaterproofIP67
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Battery1 rechargeable Lithium-ion (included)
  • Length (cm)22
  • Width (cm)9.7
  • Height (cm)6.3
  • Width (in)3 13/16
  • Length (in)8 21/32
  • Height (in)2 31/64
  • Number of Positions1
  • NIST Calibration IncludedNo
  • Probe Included5 m rugged cable gel ORP electrode
  • DescriptionPortable Dedicated pH/ORP/mV Meter with Gel ORP Electrode, 5 m Rugged Cable
  • Warranty1 year
Reviews
Q&A