Hach 5011T Sealed Glass Open-Junction pH/ATC Electrode
Representative image only.
Hach 5011T Sealed Glass Open-Junction pH/ATC Electrode
Hach | Mfr: LZW5011T.97.002| Item: 5900454
A variety of electrodes for different applications
$932.66CAD / Each
$932.66CAD / Each
Product Description
Key Features
  • Different body matierals for chemical compatibility
  • Glass body, solid polymer filled electrode for dirty water samples
  • 1 year warranty
More About this Item
Choose the electrode that is most in line with the pH range of your application.
1 Year Warranty
Specifications
  • ApplicationsSamples with high solids content
  • Electrode TypeOpen annular junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteNon-refillable solid polymer
  • Min pH (pH)2
  • Max pH (pH)14
  • Accuracy0.002 pH with MM340 Meter0.002 pH with MM374 Meter0.002 pH with pH31 Meter0.02 pH with pH3 Meter
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorBanana
  • ATC Element1000Ω Pt RTD
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)130
  • Cable Length (m)1
  • DescriptionpH Electrode w/ ATC for Laboratory, Sealed, Solid Polymer, Glass, Open Annular Junction