Hach 5-EP MG-L Hardness Drop Test Kit -Total Hardness 145401
Representative image only.
Hach 5-EP MG-L Hardness Drop Test Kit -Total Hardness 145401
Hach | Mfr: 145401| Item: 0554534
Low cost and simple drop count titration method
$85.16CAD / Each
$85.16CAD / Each
Buy It Together
Current Item
Hach 5-EP MG-L  Hardness Drop Test Kit -Total Hardness 145401
Hach 5-EP MG-L Hardness Drop Test Kit -Total Hardness 145401
0554534
$85.16CAD / Each
+ + =
Total Price 1197.72 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations.
Measure total hardness as CaCO3 with a range of 20-400 mg/L.
Rugged carrying case for analysis in the field.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestHardness, total
  • Measurement Points20 to 400 mg/L
  • Reagent PlatformEDTA Powder Pillows
  • Tests Per Kit100
  • DescriptionTotal Hardness Drop Test Kit