Hach 27426-45 Colorimeter Test Kit , Total PHosPHorus (Usepa), 0 to 3.5 ppm, 50/Kit
Representative image only.
Hach 27426-45 Colorimeter Test Kit , Total PHosPHorus (Usepa), 0 to 3.5 ppm, 50/Kit
Hach | Mfr: 2742645| Item: 9957418
Environmental test kits for your Hach Colorimeter
$185.65CAD / Each
$185.65CAD / Each
Buy It Together
Current Item
Hach 27426-45 Colorimeter Test Kit , Total PHosPHorus (Usepa), 0 to 3.5 ppm, 50/Kit
Hach 27426-45 Colorimeter Test Kit , Total PHosPHorus (Usepa), 0 to 3.5 ppm, 50/Kit
9957418
$185.65CAD / Each
+ + =
Total Price 1346.90 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Order the test kits for the Hach colorimeter to complete your water testing protocol. All kits require a colorimeter (99573-00 shown below). Chemical oxygen demand (COD), Total Nitrogen and Totol Phosphorus require a COD reactor (99574-60, 120 VAC or 99574-65, 240 VAC).
Requires COD reactor listed below.
1 Year Warranty
Specifications
  • Test ParametersTotal Phosphorus (USEPA)
  • Number of Tests Included50
  • Range0 to 3.5 ppm
  • DescriptionColorimeter Test Kit , Total PHosPHorus (Usepa), 0 to 3.5 ppm, 50/Kit