Hach 2483600 Phenol Color Disc Test Kit , 5 ppm Max, 100 Tests/Kit
Representative image only.
Hach 2483600 Phenol Color Disc Test Kit , 5 ppm Max, 100 Tests/Kit
Hach | Mfr: 2483600| Item: 0554559
Color wheel method provides accurate visual comparisons
$384.55CAD / Each
$384.55CAD / Each
Buy It Together
Current Item
Hach 2483600 PHenol Color Disc Test Kit , 5 ppm Max, 100 Tests/Kit
Hach 2483600 PHenol Color Disc Test Kit , 5 ppm Max, 100 Tests/Kit
0554559
$384.55CAD / Each
+ + =
Total Price 1545.80 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestPhenol
  • Measurement Points0 - 4.0 mg/L
  • Tests Per Kit100
  • DescriptionPHenol Color Disc Test Kit , 5 ppm Max, 100 Tests/Kit