Hach 2444400 Chlorine Drop Test Kit , High Range
Representative image only.
Hach 2444400 Chlorine Drop Test Kit , High Range
Hach | Mfr: 2444400| Item: 0554520
Low cost and simple drop count titration method
Call For Price
Call For Price
Buy It Together
Current Item
Hach 2444400 Chlorine Drop Test Kit , High Range
Hach 2444400 Chlorine Drop Test Kit , High Range
0554520
Call For Price
+ + =
Total Price 1112.56 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations.
Rugged carrying case for analysis in the field
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChlorine, high range
  • Measurement Points10 to 200 ppm
  • Reagent PlatformSulfamic Acid, KI PPs, Thiosulfate
  • Tests Per Kit100
  • DescriptionChlorine Drop Test Kit , High Range