Hach 2444301 Alkalinity Drop Test Kit -Detection (Range 5 to 100; 20 to 400 ppm)

Representative image only.

Hach 2444301 Alkalinity Drop Test Kit -Detection (Range 5 to 100; 20 to 400 ppm)

Hach | Mfr: 2444301| Item: 0554506

Low cost and simple drop count titration method

$129.81CAD / Each

$129.81CAD / Each

Buy It Together

Current Item

Hach 2444301 Alkalinity Drop Test Kit -Detection (Range 5 to 100; 20 to 400 ppm)
Hach 2444301 Alkalinity Drop Test Kit -Detection (Range 5 to 100; 20 to 400 ppm)
0554506

$129.81CAD / Each
+ + =
Total Price 1242.37 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Kit contains all required reagents and apparatus
  • 1 year warranty

More About this Item

Drop count titration is a simple method to estimate concentrations.

Rugged carrying case for analysis in the field

ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestAlkalinity, detection
  • Measurement Points5 to 100 ppm; 20 to 400 ppm
  • Reagent PlatformSulfuric Acid
  • Tests Per Kit100
  • DescriptionAlkalinity Drop Test Kit -Detection (Range 5 to 100; 20 to 400 ppm)
  • Warranty1 year
Reviews
Q&A