Hach 2444300 Alkalinity Drop Test Kit , Low Range
Representative image only.
Hach 2444300 Alkalinity Drop Test Kit , Low Range
Hach | Mfr: 2444300| Item: 0554504
Low cost and simple drop count titration method
$129.81CAD / Each
$129.81CAD / Each
Buy It Together
Current Item
Hach 2444300 Alkalinity Drop Test Kit , Low Range
Hach 2444300 Alkalinity Drop Test Kit , Low Range
0554504
$129.81CAD / Each
+ + =
Total Price 1242.37 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations.
Rugged carrying case for analysis in the field
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestAlkalinity, low range
  • Measurement Points0.4 to 8 ppm; 1 to 20 gpg
  • Reagent PlatformSulfuric Acid
  • Tests Per Kit100
  • DescriptionAlkalinity Drop Test Kit , Low Range