Hach 2428700 Nitrogen, Ammonia Color Disc Test Kit
Representative image only.
Hach 2428700 Nitrogen, Ammonia Color Disc Test Kit
Hach | Mfr: 2428700| Item: 0554575
Color wheel method provides accurate visual comparisons
$253.42CAD / Each
$253.42CAD / Each
Buy It Together
Current Item
Hach 2428700 Nitrogen, Ammonia Color Disc Test Kit
Hach 2428700 Nitrogen, Ammonia Color Disc Test Kit
0554575
$253.42CAD / Each
+ + =
Total Price 1414.67 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestNitrogen, ammonia
  • Measurement Points 0.1 to 2.5 ppm
  • Reagent PlatformSalicylate
  • Tests Per Kit100
  • DescriptionNitrogen, Ammonia Color Disc Test Kit