Hach 2359300 Molybdate Color Disc Test Kit , 3 ppm Max, 100 Tests/Kit
Representative image only.
Hach 2359300 Molybdate Color Disc Test Kit , 3 ppm Max, 100 Tests/Kit
Hach | Mfr: 2359300| Item: 0554571
Color wheel method provides accurate visual comparisons
$360.98CAD / Each
$360.98CAD / Each
Buy It Together
Current Item
Hach 2359300 Molybdate Color Disc Test Kit , 3 ppm Max, 100 Tests/Kit
Hach 2359300 Molybdate Color Disc Test Kit , 3 ppm Max, 100 Tests/Kit
0554571
$360.98CAD / Each
+ + =
Total Price 1522.23 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestMolybdate
  • Measurement Points 0.25 to 3 ppm
  • Tests Per Kit100
  • DescriptionMolybdate Color Disc Test Kit , 3 ppm Max, 100 Tests/Kit