Hach 2255000 Silica, Low Range Color Disc Test Kit
Representative image only.
Hach 2255000 Silica, Low Range Color Disc Test Kit
Hach | Mfr: 2255000| Item: 0554549
Color wheel method provides accurate visual comparisons
$324.15CAD / Each
$324.15CAD / Each
Buy It Together
Current Item
Hach 2255000 Silica, Low Range Color Disc Test Kit
Hach 2255000 Silica, Low Range Color Disc Test Kit
0554549
$324.15CAD / Each
+ + =
Total Price 1485.40 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestSilica, low range
  • Measurement Points 0.02 to 1.0 ppm
  • Tests Per Kit100
  • DescriptionSilica, Low Range Color Disc Test Kit