Hach 224100 Nitrogen Ammonia Color Disc Test Kit
Representative image only.
Hach 224100 Nitrogen Ammonia Color Disc Test Kit
Hach | Mfr: 224100| Item: 0554573
Color wheel method provides accurate visual comparisons
$235.74CAD / Each
$235.74CAD / Each
Buy It Together
Current Item
Hach 224100 Nitrogen Ammonia Color Disc Test Kit
Hach 224100 Nitrogen Ammonia Color Disc Test Kit
0554573
$235.74CAD / Each
+ + =
Total Price 1396.99 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestNitrogen, ammonia
  • Measurement Points 0.1 to 3 ppm
  • Reagent PlatformNessler Reagent
  • Tests Per Kit100
  • DescriptionNitrogen Ammonia Color Disc Test Kit