Hach 224000 Nitrate, Nitrite High Range Color Disc Test Kit
Representative image only.
Hach 224000 Nitrate, Nitrite High Range Color Disc Test Kit
Hach | Mfr: 224000| Item: 0554583
Color wheel method provides accurate visual comparisons
$328.57CAD / Each
$328.57CAD / Each
Buy It Together
Current Item
Hach 224000 Nitrate, Nitrite High Range Color Disc Test Kit
Hach 224000 Nitrate, Nitrite High Range Color Disc Test Kit
0554583
$328.57CAD / Each
+ + =
Total Price 1489.82 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestNitrite, high range
  • Measurement Points0 to 100 ppm; 40 to 2000 ppm
  • Reagent PlatformFerrous Sulfate
  • Tests Per Kit100
  • DescriptionNitrate, Nitrite High Range Color Disc Test Kit