Hach 222702 Chromium Drop Test Kit , High Range (Tests/Kit 100)
Representative image only.
Hach 222702 Chromium Drop Test Kit , High Range (Tests/Kit 100)
Hach | Mfr: 222702| Item: 0554522
Low cost and simple drop count titration method
$188.59CAD / Each
$188.59CAD / Each
Buy It Together
Current Item
Hach 222702 Chromium Drop Test Kit , High Range (Tests/Kit 100)
Hach 222702 Chromium Drop Test Kit , High Range (Tests/Kit 100)
0554522
$188.59CAD / Each
+ + =
Total Price 1301.15 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations.
Rugged carrying case for analysis in the field
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChromium, high range
  • Measurement Points5 to 100 ppm; 50 to 1000 ppm
  • Reagent PlatformThiosullfate
  • Tests Per Kit100
  • DescriptionChromium Drop Test Kit , High Range (Tests/Kit 100)