Hach 208601 Chloride Drop Test Kit , High Range
Representative image only.
Hach 208601 Chloride Drop Test Kit , High Range
Hach | Mfr: 208601| Item: 0554516
Low cost and simple drop count titration method
$321.20CAD / Each
$321.20CAD / Each
Buy It Together
Current Item
Hach 208601 Chloride Drop Test Kit , High Range
Hach 208601 Chloride Drop Test Kit , High Range
0554516
$321.20CAD / Each
+ + =
Total Price 1433.76 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations.
Rugged carrying case for analysis in the field
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChloride, high range
  • Measurement Points500 to 10,000 ppm; 5000 to 100,000 ppm
  • Reagent PlatformSilver Nitrate
  • Tests Per Kit100
  • DescriptionChloride Drop Test Kit , High Range