Hach 16900-01 Manual-Digital Titrator, without cartridges
Representative image only.
Hach 16900-01 Manual-Digital Titrator, without cartridges
Hach | Mfr: 1690001| Item: 2490800
Faster and more accurate than a conventional burette
$509.79CAD / Each
$509.79CAD / Each
Buy It Together
Current Item
Hach 16900-01 Manual-Digital Titrator, without cartridges
Hach 16900-01 Manual-Digital Titrator, without cartridges
2490800
$509.79CAD / Each
+ + =
Total Price 590.45 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
plastic case and five straight delivery tubes. Order interchangeable titrator cartridges separately.
Key Features
  • Portable, precision dispensing with automatic calculation of titrant - works in field or lab settings
  • Interchangeable, sealed cartridges with concentrated titrants save time and eliminate glassware cleanup
  • Limited Lifetime Warranty
More About this Item
The Hach digital titrator produces repeatable results through the use of concentrated titrants packaged in compact interchangeable cartridges. To titrate, simply attach a cartridge to the titrator and insert a delivery tube. Each turn of the delivery knob dispenses a calculated amount of titrant. The numerical counter records the volume. When the end point is reached, calculate the sample concentration using the number displayed on the counter. Any remaining reagent may be capped and stored for future use.
ISO9001:2000 Certified SupplierLimited Lifetime Warranty
Specifications
  • pH Accuracy± 1%
  • Dispensing Accuracy800 digits/mL or 0.00125 mL/digit
  • DescriptionManual-Digital Titrator, without cartridges
  • WarrantyLTD Life Time