Hach 1454202 Total Chlorine Test Kit , Range 0.02 to 0.68 and 0.02 to 3.4 ppm

Representative image only.

Hach 1454202 Total Chlorine Test Kit , Range 0.02 to 0.68 and 0.02 to 3.4 ppm

Hach | Mfr: 1454202| Item: 0554570

Color wheel method provides accurate visual comparisons

$228.38CAD / Each

$228.38CAD / Each

Buy It Together

Current Item

Hach 1454202 Total Chlorine Test Kit , Range 0.02 to 0.68 and 0.02 to 3.4 ppm
Hach 1454202 Total Chlorine Test Kit , Range 0.02 to 0.68 and 0.02 to 3.4 ppm
0554570

$228.38CAD / Each
+ + =
Total Price 1389.63 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty

More About this Item

Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.

ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChlorine, total
  • Measurement Points0.02 to 0.68 ppm; 0.02 to 3.4 ppm
  • Reagent PlatformDPD Method
  • Tests Per Kit100
  • DescriptionTotal Chlorine Test Kit , Range 0.02 to 0.68 and 0.02 to 3.4 ppm
  • Warranty1 year
Reviews
Q&A