Hach 1454200 Chlorine, Free and Total Color Disc Test Kit
Representative image only.
Hach 1454200 Chlorine, Free and Total Color Disc Test Kit
Hach | Mfr: 1454200| Item: 0554566
Color wheel method provides accurate visual comparisons
$218.06CAD / Each
$218.06CAD / Each
Buy It Together
Current Item
Hach 1454200 Chlorine, Free and Total Color Disc Test Kit
Hach 1454200 Chlorine, Free and Total Color Disc Test Kit
0554566
$218.06CAD / Each
+ + =
Total Price 1379.31 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestChlorine, free and total
  • Measurement Points0.02 to 0.7 ppm; 0 to 3.4 ppm
  • Reagent PlatformDPD Method
  • Tests Per Kit200
  • DescriptionChlorine, Free and Total Color Disc Test Kit