Hach 143601 Carbon Dioxide Drop Test Kit ( 1.25 to 25; 5 to 100 ppm)
Representative image only.
Hach 143601 Carbon Dioxide Drop Test Kit ( 1.25 to 25; 5 to 100 ppm)
Hach | Mfr: 143601| Item: 0554512
Low cost and simple drop count titration method
$119.64CAD / Each
$119.64CAD / Each
Buy It Together
Current Item
Hach 143601 Carbon Dioxide Drop Test Kit ( 1.25 to 25; 5 to 100 ppm)
Hach 143601 Carbon Dioxide Drop Test Kit ( 1.25 to 25; 5 to 100 ppm)
0554512
$119.64CAD / Each
+ + =
Total Price 1232.20 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations.
Rugged carrying case for analysis in the field
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestCarbon dioxide
  • Measurement Points1.25 to 25; 5 to 100 ppm
  • Reagent PlatformSodium hydroxide, Phenolphthalein
  • Tests Per Kit100
  • DescriptionCarbon Dioxide Drop Test Kit ( 1.25 to 25; 5 to 100 ppm)