Hach 1419301 Molybdate, High Range Color Disc Test Kit
Representative image only.
Hach 1419301 Molybdate, High Range Color Disc Test Kit
Hach | Mfr: 1419301| Item: 0554569
Color wheel method provides accurate visual comparisons
$543.68CAD / Each
$543.68CAD / Each
Buy It Together
Current Item
Hach 1419301 Molybdate, High Range Color Disc Test Kit
Hach 1419301 Molybdate, High Range Color Disc Test Kit
0554569
$543.68CAD / Each
+ + =
Total Price 1704.93 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestMolybdate, high range
  • Measurement Points 0.2 to 10 ppm; 1 to 50 ppm
  • Reagent PlatformMercaptoacetic Acid
  • Tests Per Kit100
  • DescriptionMolybdate, High Range Color Disc Test Kit