Hach 1408100 Nitrate, Nitrite Color Disc Test Kit
Representative image only.
Hach 1408100 Nitrate, Nitrite Color Disc Test Kit
Hach | Mfr: 1408100| Item: 0554581
Color wheel method provides accurate visual comparisons
$393.40CAD / Each
$393.40CAD / Each
Buy It Together
Current Item
Hach 1408100 Nitrate, Nitrite Color Disc Test Kit
Hach 1408100 Nitrate, Nitrite Color Disc Test Kit
0554581
$393.40CAD / Each
+ + =
Total Price 1554.65 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestNitrate, nitrite
  • Measurement Points1 to 40 ppm; 0.01 to 0.40 ppm
  • Reagent PlatformDiazotization Cadmium Reduction
  • Tests Per Kit100
  • DescriptionNitrate, Nitrite Color Disc Test Kit