GVS Polywipe Sponge for Recovery of Microorganisms on a Surface; 50/PK

Representative image only.

GVS Polywipe Sponge for Recovery of Microorganisms on a Surface; 50/PK

| Mfr: 10498521| Item: 7890951

Premoistened with neutralizing buffer to neutralize the effects of surface disinfectants

$780.90CAD / Pkg of 50

$780.90CAD / Pkg of 50
Product Description

Key Features

  • Free of preservatives that can inhibit micro-organism growth
  • Biocide free
  • Individually wrapped to maintain sterility
  • 1 year warranty

More About this Item

Easily recover microorganisms from a surface with a Polywipe sponge. The ready-to-use bright blue sponge comes premoistened with a neutralizing buffer and is free of preservatives found in commercially available sponges, which can inhibit microorganism growth. Simply wipe a surface to be analyzed then wring fluid onto a plate for analysis. Each sponge comes individually sealed in an easy-to-open pouch and is gamma irradiated to ensure sterility.

1 Year WarrantySterile
Specifications
  • MaterialSponge
  • ColorBlue
  • Length (InchShort)3 7/8
  • Height (in)0.1875
  • Width (InchShort)1 3/4
  • Width (cm)4.5
  • Height (cm)0.4
  • Length (cm)9.8
  • DescriptionPolywipe Sponge for Recovery of Microorganisms on a Surface; 50/PK
  • Warranty1 year
Reviews
Q&A