GVS Dilution Bottle, Phosphate Buffer with Magnesium Chloride, 99 mL; 72/PK
Representative image only.
GVS Dilution Bottle, Phosphate Buffer with Magnesium Chloride, 99 mL; 72/PK
| Mfr: 10498505| Item: 3615319
Eliminate the time and money spent on preparing your own dilution vials
$388.24CAD / Pkg of 72
BUY MORE & SAVE
  • 1-4$388.24 CAD / Pkg of 72
  • 5-6$349.42 CAD / Pkg of 72
  • 7-9$330.00 CAD / Pkg of 72
  • 10+$310.59 CAD / Pkg of 72
$388.24CAD / Pkg of 72
BUY MORE & SAVE
  • 1-4$388.24 CAD / Pkg of 72
  • 5-6$349.42 CAD / Pkg of 72
  • 7-9$330.00 CAD / Pkg of 72
  • 10+$310.59 CAD / Pkg of 72
Buy It Together
Current Item
GVS Dilution Bottle, Phosphate Buffer with Magnesium Chloride, 99 mL; 72/PK
GVS Dilution Bottle, Phosphate Buffer with Magnesium Chloride, 99 mL; 72/PK
3615319
$388.24CAD / Pkg of 72
+ =
Total Price 388.24 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Prefilled bottles make sample preparation easy and consistent
  • Final pH for all solutions is 7.2 pH ±0.2 pH at 25°C
  • Sterility makes these suitable for sensitive microbiological and pharmaceutical processes
  • 1 year warranty
More About this Item
Ensure consistent dilutions and reduce the risk of contamination with these prefilled, sterile dilution bottles. Designed with an easy open, flip-top, plastic container with tamper-evident seal to produce an accurate and safe sample dilution. These buffers are ideal for sample dilution of water, dairy products, foods and pharmaceuticals prior to microbiological testing. Choose from Butterfield’s Phosphate Buffer or Phosphate Buffer with Magnesium Chloride.
Butterfield’s Buffer contains monobasic potassium phosphate and is used extensively in the food, dairy, and pharmaceutical industries to ensure the stable pH during the testing process and does not interfere with microbial growth. Offered in 90 ml and 99 ml volumes for easy 1:10 and 1:100 dilutions. Butterfield’s buffer is recommended as a general diluent in laboratory procedures by the Federal Drug Administrations and in the Bacteriological Analytical Manual. It is recommended by APHA for the recovery of injured microorganisms from dairy and food samples.
Phosphate Buffer with magnesium chloride is used as the diluents for the preparation of dilutions in plate counts in the dairy and food industries. This product is prepared according to Standard Methods for the Examination of Water and Wastewater for use in water testing. It Contains deionized water, monopotassium phosphate, and magnesium chloride.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Sterile
Specifications
  • Element/IonPhosphate Buffer with Magnesium Chloride
  • FormLiquid
  • Volume (mL)99
  • PackagingFlip-top Plastic Bottle with Tamper-evident Seal
  • DOT InformationNot regulated
  • Shelf Life (months)12
  • DescriptionDilution Bottle, Phosphate Buffer with Magnesium Chloride, 99 mL; 72/PK