Guardian Equipment G1101 Faucet-Mount Personal Eyewash with gooseneck-mounted outlets
Representative image only.
Guardian Equipment G1101 Faucet-Mount Personal Eyewash with gooseneck-mounted outlets
Guardian Equipment | Mfr: G1101| Item: 0676744
Installs easily on standard faucet; water pressure keeps stream flowing
$289.89CAD / Each
$289.89CAD / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Adjustable outlets are mounted on bends 5" (12.7 cm) apart and angled inward. Chrome-plated brass construction.
Compliance: ANSI Z358.1-2004 as personal eyewash only.
Two year warranty
Specifications
  • Eyewash Fixture MaterialChrome-plated brass
  • Supply Pipe MaterialChrome-plated brass
  • MountFaucet Mount
  • Inlet Connection55/64"-27 (F) for faucet
  • DescriptionFaucet-Mount Personal Eyewash with gooseneck-mounted outlets