Foxx Life Sciences UN/DOT Liquid Waste Container, S70 cap; 13.5 L; 1/ea

Representative image only.

Foxx Life Sciences UN/DOT Liquid Waste Container, S70 cap; 13.5 L; 1/ea

| Mfr: 392-2003-OEM| Item: 0605998

Safely transport hazardous liquids

$54.66CAD / Each

$54.66CAD / Each

Buy It Together

Current Item

Foxx Life Sciences UN/DOT Liquid Waste Container, S70 cap; 13.5 L; 1/ea
Foxx Life Sciences UN/DOT Liquid Waste Container, S70 cap; 13.5 L; 1/ea
0605998

$54.66CAD / Each
+ =
Total Price 320.31 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

70-mm HDPE cap.

Key Features

  • UN/DOT rated for Group II and hazardous chemicals
  • Large top handle provides a comfortable full-hand grip
  • HDPE construction offers excellent chemical compatibility

More About this Item

Sturdy, rectangular UN/DOT carboys are easy to fill and ship for disposal. Safely use these single-use carboys to transfer and store liquid waste, use as waste traps with automated instruments, or as part of your HPLC system.

UN rating: 3H1/Y1.8/100

ISO9001 Registered Supplier
Specifications
  • Capacity (L)13.5
  • MaterialHDPE
  • Cap MaterialHDPE
  • Cap Size (mm)70
  • Overall Length (in)11
  • Overall Width (in)9
  • Overall Height (in)11
  • DescriptionWaste Container, UN/DOT, S70 cap; 13.5 L; 1/ea
Reviews
Q&A