Fowler 54-115-100-0 Wireless Transmitter for Caliper/Indicator
Representative image only.
Fowler 54-115-100-0 Wireless Transmitter for Caliper/Indicator
Fowler | Mfr: 54-115-100-0| Item: 0992549
Wirelessly transfer data from your calipers
$349.19CAD / Each
$349.19CAD / Each
Product Description
Includes
USB receiver, transmitter (good for 1 tool), spare battery (CR2032), and storage case.
Key Features
  • Compatible with digital calipers 97152-06 and -08
  • Use of the tool up to 60 feet away from a computer
  • Audible as well as LED indications when data is sent
  • Input into Excel, Word, Notepad and CAD programs with the press of a button
  • Simple plug and play operation
More About this Item
Wireless transmitter and USB receiver to wirelessly transmit measurement data to your PC.
Conformite Europeenne
Specifications
  • Compatible WithDigital calipers 97152-06 and -08
  • DescriptionWireless Transmitter for Caliper/Indicator